Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
β-L-2'-Deoxyadenosine
β-L-2'-Deoxyadenosine ≥98% (by HPLC) MORpure™
β-L-2'-Deoxyadenosine ≥98% (by HPLC) MORpure™
β-L-2'-Deoxyadenosine ≥98% (by HPLC) MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
β-L-2'-Deoxyadenosine ≥98% (by HPLC) MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
14365-45-8
: