Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
β-L-2'-Deoxyguanosine
β-L-2'-Deoxyguanosine ≥98% (by HPLC) MORpure™
β-L-2'-Deoxyguanosine ≥98% (by HPLC) MORpure™
β-L-2'-Deoxyguanosine ≥98% (by HPLC) MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
Certificates
Expand image
β-L-2'-Deoxyguanosine ≥98% (by HPLC) MORpure™
Supplier:
MORAVEK BIOCHEMICALS MS
About this item
CAS Number:
22837-44-1
:
Formula:
C₁₀H₁₃N₅O₄
MDL Number:
MFCD07367081
:
267.241 g/mol