Clevudine ≥98% (by HPLC) MORpure™
Catalog # 101969-804
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial, with screwcap
- Size:1 mg
- Cat. No.:101969-804
- Supplier no.:M1885-1MG
Specifications
About this item
:
:260.2200 g/mol
CAS Number:163252-36-6
Formula:C₁₀H₁₃FN₂O₅
MDL Number:MFCD00935785