8-Bromo-1H-purine-2,6(3H,7H)-dione
Catalog # 101969-242
Supplier: MORAVEK BIOCHEMICALS MS
CAS Number:
Specifications
- Pack type:NextGen V-Vial, with screwcap
- Size:1 mg
- Cat. No.:101969-242
- Supplier no.:M1399-1MG
Specifications
About this item
:
:231.01 g/mol
CAS Number:10357-68-3
Formula:C₅H₃BrN₄O₂
MDL Number:MFCD02683597