Order Entry
Puerto Rico
ContactUsLinkComponent
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-methyluracil, [uracil-2-14C] ≥98% (by HPLC), MORpure™, HPLC purified
Irritant
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-methyluracil, [uracil-2-14C] ≥98% (by HPLC), MORpure™, HPLC purified
Catalog # 101970-964
CAS Number:  
undefined
Irritant
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-methyluracil, [uracil-2-14C] ≥98% (by HPLC), MORpure™, HPLC purified
Catalog # 101970-964
Supplier Number:  MC2041-50UCI
CAS Number:  

Specifications

  • Pack type:
    NextGen V-Vial
  • Size:
    50 µCi
  • Cat. No.:
    101970-964
  • Supplier no.:
    MC2041-50UCI

Specifications

About this item

 :
UN:2910
ADR:,
 :GHS07
CAS Number:83374-60-1