ContactUsLinkComponent
OilExpress Flow Cell, SP100, PerkinElmer
OilExpress Flow Cell, SP100, PerkinElmer
Catalog # 10133-724
Supplier:  PerkinElmer
OilExpress Flow Cell, SP100, PerkinElmer
Catalog # 10133-724
Supplier:  PerkinElmer
Supplier Number:  L1250220
Specifications
  • Description:
    OilExpress Flowcell, 0.1 mm pathlength
  • Cat. No.:
    10133-724
  • Supplier no.:
    L1250220
Specifications
About this item
OilExpress Flowcell with wedged Spacer, ZnSe Windows (one grooved), and two ports.