Inosine 5'-monophosphate disodium salt hydrate ≥98% (by HPLC), crystalline powder
Supplier: Chem Impex International
New Product
Certificates
About this item
Synonyms: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate
CAS Number:352195-40-5
Formula:C₁₀H₁₁N₄Na₂O₈P·xH₂O
MDL Number:MFCD00150372
:392.17 g/mol (anhy)
Storage Temperature:Refrigerator
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White or off-white crystalline powder
- Heavy Metals:≤ 10 ppm
- Arsenic:≤ 1 ppm
- Lead :≤ 1.5 ppm
- Water Content (Karl Fisher):≤ 26%
- pH:7.0 - 8.5
- Yeast and Mold:≤ 30 cfu/g
- Coliform Organisms:Absence/10g
- E. Coli:Absent
- Microbial Count:≤ 600 cfu/g (TPC)
- Salmonella:Absent
- Purity by HPLC:≥ 98%