Soybean Bile Broth Base, Modified, Dehydrated Culture Media, HiMedia Laboratories
Catalog # 95020-870
Supplier: HiMedia
CAS Number:
Specifications
- Description:Soybean Bile Broth Base, Modified
- Cat. No.:95020-870
- Supplier no.:M1286I-500G
Specifications
About this item
Recommended as an enrichment medium for the detection of Escherichia coli O157:H7 from food and animal foodstuffs.
- Microbiological culture media
- 500 g poly bottle