Orders LinkContactUsLinkComponent
Open Handle Probes, Seedburo
Open Handle Probes, Seedburo
Supplier:   Seedburo Equipment
Open Handle Probes, Seedburo
Supplier:  Seedburo Equipment
Certificates
Open Handle Probes, Seedburo
Open Handle Probes, Seedburo
2 Options Available Below
Product Details & Documents
Open-ended probes without partitions are more convenient to empty and are widely used by the grain elevator operators for obtaining samples from truck loads.
  • Extra heavy gauge tubing for durable, time-tested sampling
  • Held upright and sample is dumped from the handle
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 56724-039
    Open Handle Probe, Brass, 10 openings
    Open Handle Probe, Brass, 10 openings
    160 cm (63")
    Log in for availability
    Log in for availability
    Each
    Each - $1,590.00
    $1,590.00
    Each
       $1,590.00
    Minimum order quantity is 1
    Supplier Number: 22-OH
  • 56724-038
    Open Handle Probe, Brass, 8 openings
    Open Handle Probe, Brass, 8 openings
    129.5 cm (51")
    Log in for availability
    Log in for availability
    Each
    Each - $1,043.37
    $1,043.37
    Each
       $1,043.37
    Minimum order quantity is 1
    Supplier Number: 21-OH