BD Taxo™ TB Niacin Test Control, BD Diagnostic Systems
BD231741
: BD
:
- Pack type:Dispensing Cartridge
- Description:BD Taxo™ TB niacin test strips
- Size:25 strips
- Cat. No.:BD231741
- Supplier no.:231741
BD Taxo™ TB Niacin Test Controls are impregnated with nicotinamide and give a positive reaction when tested with a TB Niacin Test Strip.