

Acetonitrile with 0,1% (v/v) formic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
:75-05-8
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:−45,7 °C
Storage Temperature:Ambient
UN:1648
ADR:3,
:M-Clarity™ quality level = MQ100
Flash Pt:2 °C
:GHS02
:GHS07
Frequently Bought Together