Potassium dihydrogen phosphate ≥99.5% for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Melting Pt:253 °C
Density:2.34 g/cm³ (20 °C) (lit.)
Storage Temperature:Ambient
EINECS:231-913-4
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay:99.5 - 101.0 %
- pH (10 %, 20°C):4.1 - 4.5
- Absorbance at 210 nm:Max. 0.1
- Absorbance at 220 nm:Max. 0.06
- Absorbance at 230 nm:Max. 0.04
- Absorbance at 300 nm:Max. 0.02
- Absorbance 500 nm:Max. 0.02
- Appearance of the solution (c=10%):complying
- pKa (25 °C):(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)