Order Entry
Puerto Rico
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) stabilized horseradish peroxidase (HRP) substrate
3,3',5,5'-Tetramethylbenzidine (TMB) stabilized horseradish peroxidase (HRP) substrate
Catalog # PAW4121
CAS Number:  
undefined
3,3',5,5'-Tetramethylbenzidine (TMB) stabilized horseradish peroxidase (HRP) substrate
Catalog # PAW4121
Supplier Number:  W4121
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    200 ml
  • Cat. No.:
    PAW4121
  • Supplier no.:
    W4121

Specifications

About this item

 :
 :240.35 g/mol
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748