Cleanroom Shoe Covers, Veltek
Supplier: Veltek Associates
Certificates
About this item
These Cleanroom shoe covers are made of ISO Class: 5 special polyolefin blend laminated to a polypropylene substrate.
- Durable, resistant to liquids, non-skid soles
- Elasticized top opening
- Excellent skid resistance
Specifications
- Catalog No:
- 75795-588
- 75795-420
- Garment application:
- Cleanroom
- Cleanroom