Analytical Funnels
About this item
GVS analytical funnels with white/black grids are supplied sterile.
- Ready-to-use filtration unit
- Pre-sterilized
- No flaming
- Minimizes the risk of cross-contamination
- Easy release of membrane
GVS microbiological monitors and analytical funnels provide a complete system solution for liquid sample preparation. Each single-use, pre-sterilized filtering unit consists of a measured filter funnel, base, pad, membrane, removable lid and plug. This all-in-one system easily converts from the 100 ml filtration unit to a petri dish, which can be labeled and incubated for culturing. The GVS funnels meet the standard method requirements for a disposable device.
6 Options Available Below
Product Details & Documents
GVS analytical funnels with white/black grids are supplied sterile.
- Ready-to-use filtration unit
- Pre-sterilized
- No flaming
- Minimizes the risk of cross-contamination
- Easy release of membrane
GVS microbiological monitors and analytical funnels provide a complete system solution for liquid sample preparation. Each single-use, pre-sterilized filtering unit consists of a measured filter funnel, base, pad, membrane, removable lid and plug. This all-in-one system easily converts from the 100 ml filtration unit to a petri dish, which can be labeled and incubated for culturing. The GVS funnels meet the standard method requirements for a disposable device.
Product Family Options
Product Information
- MaterialPore sizeMembrane diameterAvailabilityPrice
- 10057-706Analytical Funnel, White/Black GridAnalytical Funnel, White/Black GridNitrocellulose0.20 μm47 mmLog in for availabilityPack of 50$468.95Supplier Number: 10497507Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 10057-708Analytical Funnel, White/Black GridAnalytical Funnel, White/Black GridNitrocellulose0.45 μm47 mmLog in for availabilityPack of 50$468.95Supplier Number: 10497504Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 10057-704Analytical Funnel, White/Black Grid, Individually baggedAnalytical Funnel, White/Black Grid, Individually baggedNitrocellulose0.20 μm47 mmLog in for availabilityPack of 50$458.61Supplier Number: 10497510Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 10057-710Analytical Funnel, White/Black Grid, Individually baggedAnalytical Funnel, White/Black Grid, Individually baggedNitrocellulose0.45 μm47 mmLog in for availabilityPack of 50$458.61Supplier Number: 10497506Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 10057-774Analytical Funnel, Black/White GridAnalytical Funnel, Black/White GridNitrocellulose0.45 μm47 mmLog in for availabilityPack of 50$468.95Supplier Number: 10497508
- 10057-776Analytical Funnel, White/Black Grid, Individually baggedAnalytical Funnel, White/Black Grid, Individually baggedNitrocellulose0.45 μm47 mmLog in for availabilityPack of 50$458.61Supplier Number: 10497509