M-Tergitol 7 Agar with Meat Extract, Dehydrated, HiMedia
Catalog # 95039-320
Supplier: HiMedia
CAS Number:
Specifications
- Description:M-Tergitol* 7 Agar with Meat Extract
- Cat. No.:95039-320
- Supplier no.:M1678-500G
Specifications
About this item
A selective and differential medium for recovery of injured coliforms from chlorinated water using membrane filter technique.
- Microbiological culture media
- 500 g poly bottles