Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114]
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114]
Supplier:  Bio X Cell
Certificates
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114]
Supplier:  Bio X Cell

Specifications

  • Catalog No:
  • 76555-100
  • 76556-816
  • 76554-552
  • 76556-894
  • 76555-662
  • Antigen Symbol:
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • MHC
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • Antigen Name:
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • M5/114
  • M5/114
  • M5/114
  • M5/114
  • M5/114
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • IgG2b
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Yes

Specifications