Anti-MHC Class I (H-2Kd) Mouse Monoclonal Antibody [clone: SF1.1.10 (HB-159)]
InVivoMab anti-mouse MHC Class I (H-2Kd), Clone: SF1.1.10 (HB-159), Isotype: mouse IgG2a, Applications: Purification of MHC peptide complexes, Flow cytometry
Type: Primary
Antigen: MHC Class I (H-2Kd)
Clonality: Monoclonal
Clone: SF1.1.10 (HB-159)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
- Catalog No:
- 76556-772
- 76555-906
- 76555-494
- 76556-286
- 76556-256
- Antigen Symbol:
- MHC Class I (H-2Kd)
- MHC Class I (H-2Kd)
- MHC Class I (H-2Kd)
- MHC
- MHC Class I (H-2Kd)
- Antigen Name:
- MHC Class I (H-2Kd)
- MHC Class I (H-2Kd)
- MHC Class I (H-2Kd)
- MHC Class I (H-2Kd)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- SF1.1.10 (HB-159)
- SF1.1.10 (HB-159)
- SF1.1.10 (HB-159)
- SF1.1.10 (HB-159)
- SF1.1.10 (HB-159)
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes