Anti-IL-2 Rat Monoclonal Antibody [clone: JES6-1A12]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-mouse IL-2, Clone: JES6-1A12, Isotype: rat IgG2a, κ, Applications: in vivo IL-2 neutralization, in vivo IL-2 receptor stimulation (as a complex with IL-2)
Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: JES6-1A12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Specifications
- Catalog No:
- 76554-580
- 76554-558
- 76554-534
- 76555-914
- 76556-856
- Antigen Symbol:
- IL-2
- IL-2
- IL-2
- IL-2
- IL-2
- Antigen Name:
- IL-2
- IL-2
- IL-2
- IL-2
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- JES6-1A12
- JES6-1A12
- JES6-1A12
- JES6-1A12
- JES6-1A12
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- Neutralisation:
- Yes
- Yes
- Yes
- Yes