Anti-MHC Class I (H-2Kb) Mouse Monoclonal Antibody [clone: Y-3]
InVivoMab anti-mouse MHC Class I (H-2Kb), Clone: Y-3, Isotype: mouse IgG2b, κ, Applications: Functional assays, Purification of MHC peptide complexes, Flow cytometry
Type: Primary
Antigen: MHC Class I (H-2Kb)
Clonality: Monoclonal
Clone: Y-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
- Catalog No:
- 76556-398
- 76556-954
- 76556-030
- 76555-920
- 76556-128
- Antigen Symbol:
- H-2Kb
- MHC
- H-2Kb
- H-2Kb
- H-2Kb
- Antigen Name:
- MHC Class I (H-2Kb)
- MHC Class I (H-2Kb)
- MHC Class I (H-2Kb)
- MHC Class I (H-2Kb)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- Y-3
- Y-3
- Y-3
- Y-3
- Y-3
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes