Anti-MHC Class II (beta chain) Hamster Monoclonal Antibody [clone: KL277]
InVivoMab anti-mouse MHC Class II (β chain), Clone: KL277, Isotype: hamster/mouse IgG, Applications: Western blot
Type: Primary
Antigen: MHC Class II (beta chain)
Clonality: Monoclonal
Clone: KL277
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 76556-736
- 76555-106
- 76555-180
- 76554-388
- 76556-126
- Antigen Symbol:
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- Antigen Name:
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- MHC Class II (beta chain)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- KL277
- KL277
- KL277
- KL277
- KL277
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Hamster
- Hamster
- Hamster
- Hamster
- Hamster
- Isotype:
- IgG
- IgG
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- Yes