Anti-MHC Class I (HLA-A, HLA-B, HLA-C) Mouse Monoclonal Antibody [clone: W6/32]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-human MHC Class I (HLA-A, HLA-B, HLA-C), Clone: W6/32, Isotype: mouse IgG2a, κ, Applications: Functional assays
Type: Primary
Antigen: MHC Class I (HLA-A, HLA-B, HLA-C)
Clonality: Monoclonal
Clone: W6/32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Specifications
- Catalog No:
- 76556-642
- 76555-156
- 76555-436
- 76556-364
- 76557-356
- Antigen Symbol:
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC
- Antigen Name:
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC Class I (HLA-A, HLA-B, HLA-C)
- MHC Class I (HLA-A, HLA-B, HLA-C)
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- W6/32
- W6/32
- W6/32
- W6/32
- W6/32
- Reactivity:
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a