ContactUsLinkComponent
BD™ FecalSwab™ Fecal Collection and Transport System, BD Diagnostics
BD™ FecalSwab™ Fecal Collection and Transport System, BD Diagnostics
Catalog # 75833-310
Supplier:  BD
BD™ FecalSwab™ Fecal Collection and Transport System, BD Diagnostics
Catalog # 75833-310
Supplier:  BD
Supplier Number:  220258
Specifications
  • Type:
    Single
  • Description:
    Regular Flocked Swab
  • Cat. No.:
    75833-310
  • Fill solution:
    FecalSwab™ Transport and Preservative Medium
  • Supplier no.:
    220258
Specifications
About this item
FecalSwab™ plastic screw-cap tube with conical shaped bottom filled with 2 mL of FecalSwab™ transport and preservative medium with one swab with flocked nylon fiber tip.
FecalSwab is a convenient system for transporting fecal samples in small instrument-ready tubes, saving space and making it easier to transport to the laboratory.