Human IgG Isotype Control [clone: CTLA-4-Ig (hum./hum.)]
InVivoPlus recombinant CTLA-4-Ig (hum./hum.), Clone: CTLA-4-Ig (hum./hum.), Isotype: human IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen: CTLA-4
Clonality: Monoclonal
Clone: CTLA-4-Ig (hum./hum.)
Conjugation:
Epitope:
Host: Human
Isotype: IgG
Reactivity:
- Catalog No:
- 76555-538
- 76555-408
- 76555-378
- 76555-998
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- CTLA-4-Ig (hum./hum.)
- CTLA-4-Ig (hum./hum.)
- CTLA-4-Ig (hum./hum.)
- CTLA-4-Ig (hum./hum.)
- Host:
- Human
- Human
- Human
- Human
- Isotype:
- IgG
- IgG
- IgG
- IgG
- Purification:
- Protein G
- Protein G
- Protein G
- Protein G
- Purity:
- >95%
- >95%
- >95%
- >95%