Anti-LFA-1alpha (CD11a) Mouse Monoclonal Antibody [clone: TS-1/22.1.1.13]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab anti-human LFA-1α (CD11a), Clone: TS-1/22.1.1.13, Isotype: mouse IgG1, Applications: in vitro LFA-1 neutralization
Type: Primary
Antigen: LFA-1alpha (CD11a)
Clonality: Monoclonal
Clone: TS-1/22.1.1.13
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 76555-770
- 76555-088
- 76556-676
- 76557-246
- 76557-052
- Antigen Symbol:
- LFA-1alpha
- LFA-1alpha
- LFA-1alpha
- LFA-1alpha
- LFA-1alpha
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- TS-1/22.1.1.13
- TS-1/22.1.1.13
- TS-1/22.1.1.13
- TS-1/22.1.1.13
- TS-1/22.1.1.13
- Reactivity:
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- IgG1
- Neutralisation:
- Yes
- Yes
- Yes
- Yes