Mouse IgG1 Isotype Control [clone:MOPC-21]
Supplier: Bio X Cell
Certificates
About this item
InVivoPlus mouse IgG1 isotype control, Clone: MOPC-21, Isotype: mouse IgG1, κ, Applications: in vivo depletion of Gr-1+ myeloid cells, Flow cytometry, Immunohistochemistry (paraffin), Immunohistochemistry (frozen)
- Recommended Dilution Buffer: IP0065
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- 76556-504
- 76555-850
- 76557-292
- 76554-326
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- MOPC-21
- MOPC-21
- MOPC-21
- MOPC-21
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- IgG1