Order Entry
Puerto Rico
ContactUsLinkComponent
Rat IgG1 Isotype Control [clone:HRPN]
Rat IgG1 Isotype Control [clone:HRPN]
Supplier:  Bio X Cell
Certificates
Rat IgG1 Isotype Control [clone:HRPN]
Supplier:  Bio X Cell

Specifications

  • Catalog No:
  • 76556-748
  • 76557-348
  • 76555-800
  • 76556-216
  • Antibody Type:
  • Control
  • Control
  • Control
  • Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • HRPN
  • HRPN
  • HRPN
  • HRPN
  • Host:
  • Rat
  • Rat
  • Rat
  • Rat
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • IgG1

Specifications