Mouse IgG2a Isotype Control [clone:C1.18.4]
InVivoPlus mouse IgG2a isotype control, Clone: C1.18.4, Isotype: mouse IgG2a, κ, Applications: in vivo depletion of Gr-1+ myeloid cells, Flow cytometry, Immunohistochemistry (paraffin), Immunohistochemistry (frozen)
- Recommended Dilution Buffer: IP0070
Type: Control
Antigen:
Clonality: Monoclonal
Clone: C1.18.4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
- Catalog No:
- 76556-728
- 76554-372
- 76556-704
- 76554-788
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- C1.18.4
- C1.18.4
- C1.18.4
- C1.18.4
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a