Rat IgG2a Isotype Control [clone:2A3]
Supplier: Bio X Cell
Certificates
About this item
InVivoPlus rat IgG2a isotype control, Clone: 2A3, Isotype: rat IgG2a, κ, Applications: in vivo administration
- Recommended Dilution Buffer: IP0065
Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Specifications
- Catalog No:
- 76555-684
- 76557-144
- 76555-708
- 76556-230
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 2A3
- 2A3
- 2A3
- 2A3
- Host:
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a