Mouse IgG Isotype Control
Supplier: Bio X Cell
About this item
InVivoMab polyclonal mouse IgG, Clone: Polyclonal, Isotype: mouse IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
InVivoMab polyclonal mouse IgG, Clone: Polyclonal, Isotype: mouse IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Specifications for Product Family
Specifications
- Catalog No:
- 103572-504
- 76555-688
- 103252-616
- 76555-466
- 103252-618
- 76556-244
- 103252-620
- 76555-738
- 103252-614
- 76554-526
- Antigen Symbol:
- IgG
- IgG
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Polyclonal
- Polyclonal
- Polyclonal
- Polyclonal
- Polyclonal
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG
- IgG
- IgG
- IgG
- IgG
- IgG
- IgG
- IgG
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 1 mgDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.103572-504Antigen SymbolIgGAntibody TypeClonalityHostMouseIsotypeMarketsource - Supplier Number: BE0093-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76555-688Antigen SymbolIgGAntibody TypeClonalityHostMouseIsotype - 5 mgDiscontinued-Supplier Number: -Specifications:Marketsource
More
Specifications
Cat. No.103252-616Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgGMarketsource - Supplier Number: BE0093-5MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76555-466Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgG - 25 mgDiscontinued-Supplier Number: -Specifications:Marketsource
More
Specifications
Cat. No.103252-618Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgGMarketsource - Supplier Number: BE0093-25MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76556-244Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgG - 50 mgDiscontinued-Supplier Number: -Specifications:Marketsource
More
Specifications
Cat. No.103252-620Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgGMarketsource - Supplier Number: BE0093-50MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76555-738Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgG - 100 mgDiscontinued-Supplier Number: -Specifications:Marketsource
More
Specifications
Cat. No.103252-614Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgGMarketsource - Supplier Number: BE0093-100MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76554-526Antigen SymbolAntibody TypePrimaryClonalityPolyclonalHostMouseIsotypeIgG



