Mouse IgG2b Isotype Control [clone: MPC-11]
Supplier: Bio X Cell
Certificates
About this item
InVivoMab mouse IgG2b isotype control, Clone: MPC-11, Isotype: mouse IgG2b, κ, Applications: in vivo administration, Functional assays, Immunohistochemistry (frozen), Immunohistochemistry (paraffin), Western blot, Immunofluorescence, Flow Cytometry
- Recommended Dilution Buffer: IP0070
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MPC-11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Specifications
- Catalog No:
- 76555-144
- 76555-904
- 76555-274
- 76554-598
- 76556-340
- Antigen Symbol:
- Mouse
- Antibody Type:
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- MPC-11
- MPC-11
- MPC-11
- MPC-11
- MPC-11
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes