Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse IgG2a Isotype Control [clone: C1.18.4]
Mouse IgG2a Isotype Control [clone: C1.18.4]
Supplier:  Bio X Cell
Certificates
Mouse IgG2a Isotype Control [clone: C1.18.4]
Supplier:  Bio X Cell

Specifications

  • Catalog No:
  • 76555-826
  • 76555-610
  • 76556-498
  • 76556-828
  • 76556-416
  • Antigen Symbol:
  • Mouse
  • Antibody Type:
  • Control
  • Control
  • Control
  • Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • C1.18.4
  • C1.18.4
  • C1.18.4
  • C1.18.4
  • C1.18.4
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a
  • IgG2a
  • IgG2a
  • IgG2a

Specifications