Anti-PRPF38A Rabbit Polyclonal Antibody
Catalog # 76076-588
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Pre-mRNA-splicing factor 38A
- Antigen Symbol:PRPF38A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:283-312 aa
- Form:Liquid
- Gene ID:84950
- Modification:None
- Molecular Weight:37 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This PR38A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 283-312 amino acids from the C-terminal region of human PR38A.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76076-588
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: PRPF38A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 283-312
Host: Rabbit
Isotype: Ig
Reactivity: Human