Anti-ACY3 Rabbit Polyclonal Antibody
Catalog # 76075-456
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)
- Antigen Symbol:ACY3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:66-95 aa
- Form:Liquid
- Gene ID:91703
- Modification:None
- Molecular Weight:35 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4°C
- Immunogen:This ACY3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 66-95 amino acids from the Central region of human ACY3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76075-456
Specifications
About this item
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: ACY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 66-95
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse