Anti-PFKFB1 Rabbit Polyclonal Antibody
Catalog # 76075-150
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
- Antigen Symbol:PFKFB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:13-43 aa
- Form:Liquid
- Gene ID:5207
- Antigen Synonyms:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3,6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase|6PF-2-K/Fru-2,6-P2ase 3,6-bisphosphatase|IPFK2|inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase|Renal carcinoma antigen NY-REN-56|FLJ37326|6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase
- Modification:None
- Molecular Weight:55 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping Temperature:4°C
- Immunogen:This PFKFB1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 13-43 amino acids from the N-terminal region of human PFKFB1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76075-150
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100
Type: Primary
Antigen: PFKFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 13-43
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse