Anti-ALDH4A1 Rabbit Polyclonal Antibody
Catalog # 76074-824
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial
- Antigen Symbol:ALDH4A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:533-561 aa
- Form:Liquid
- Gene ID:8659
- Modification:None
- Molecular Weight:62 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping Temperature:4°C
- Immunogen:This ALDH4A1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 533-561 amino acids from the C-terminal region of human ALDH4A1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76074-824
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100
Type: Primary
Antigen: ALDH4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 533-561
Host: Rabbit
Isotype: Ig
Reactivity: Human