Anti-CDC25C Rabbit Polyclonal Antibody
Catalog # 76074-760
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:M-phase inducer phosphatase 3
- Antigen Symbol:CDC25C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:376-402 aa
- Form:Liquid
- Gene ID:995
- Modification:None
- Molecular Weight:53 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping Temperature:4°C
- Immunogen:This CDC25C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 376-402 amino acids from the C-terminal region of human CDC25C.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76074-760
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: CDC25C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 376-402
Host: Rabbit
Isotype: Ig
Reactivity: Human