Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # 76073-586
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:5'-AMP-activated protein kinase catalytic subunit alpha-1
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Form:Liquid
- Gene ID:5562
- Modification:None
- Molecular Weight:64 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4°C
- Immunogen:This PRKAA1 antibody is generated from rabbits immunized with a recombinant protein from human PRKAA1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76073-586
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human