Anti-AK7 Rabbit Polyclonal Antibody
Catalog # 76073-352
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Adenylate kinase 7
- Antigen Symbol:AK7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:629-656 aa
- Form:Liquid
- Gene ID:122481
- Modification:None
- Molecular Weight:83 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping Temperature:4°C
- Immunogen:This AK7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 629-656 amino acids from the C-terminal region of human AK7.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76073-352
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100
Type: Primary
Antigen: AK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 629-656
Host: Rabbit
Isotype: Ig
Reactivity: Human