Anti-ECE1 Rabbit Polyclonal Antibody
76072-992
: Prosci
- Antibody Type:Primary
- Antigen Name:Endothelin-converting enzyme 1
- Antigen Symbol:ECE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:392-419 aa
- Form:Liquid
- Gene ID:1889
- Modification:None
- Molecular Weight:87 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.31 mg/ml
- Shipping Temperature:4°C
- Immunogen:This ECE-1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 392-419 amino acids from the Central region of human ECE-1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76072-992
For WB starting dilution is: 1:1000
Type: Primary
Antigen: ECE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 392-419
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse