Anti-ACADM Rabbit Polyclonal Antibody
Catalog # 76072-958
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Medium-chain specific acyl-CoA dehydrogenase, mitochondrial
- Antigen Symbol:ACADM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:189-217 aa
- Form:Liquid
- Gene ID:34
- Modification:None
- Molecular Weight:47 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping Temperature:4°C
- Immunogen:This ACADM antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 189-217 amino acids from the Central region of human ACADM.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76072-958
Specifications
About this item
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100 For FACS starting dilution is: 1:10~50 For IF starting dilution is: 1:10~50
Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 189-217
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse