Anti-MTM1 Rabbit Polyclonal Antibody
Catalog # 76072-944
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Myotubularin
- Antigen Symbol:MTM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:566-594 aa
- Form:Liquid
- Gene ID:4534
- Modification:None
- Molecular Weight:70 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4°C
- Immunogen:This MTM1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 566-594 amino acids from the C-terminal region of human MTM1.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76072-944
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: MTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 566-594
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse