Anti-USP17L24 Rabbit Polyclonal Antibody
Catalog # 76071-856
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 17-like protein 24
- Antigen Symbol:USP17L24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:496-524 aa
- Form:Liquid
- Gene ID:728369, 728373, 728379, 728393, 728400, 728405, 728419
- Modification:None
- Molecular Weight:60 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This USP17 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 496-524 amino acids from the C-terminal region of human USP17.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76071-856
Specifications
About this item
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: USP17L24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 496-524
Host: Rabbit
Isotype: Ig
Reactivity: Human