Anti-ATF6B Rabbit Polyclonal Antibody
Catalog # 76071-538
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclic AMP-dependent transcription factor ATF-6 beta
- Antigen Symbol:ATF6B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Hamster,Human,Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:560-589 aa
- Form:Liquid
- Gene ID:1388
- Modification:None
- Molecular Weight:77 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This CREBL1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 560-589 amino acids from the C-terminal region of human CREBL1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76071-538
Specifications
About this item
For IHC-P starting dilution is: 1:25 For WB starting dilution is: 1:1000
Type: Primary
Antigen: ATF6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 560-589
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Hamster