Anti-ATF1 Rabbit Polyclonal Antibody
76071-346
: Prosci
- Antibody Type:Primary
- Antigen Name:Cyclic AMP-dependent transcription factor ATF-1
- Antigen Symbol:ATF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:193-219 aa
- Form:Liquid
- Gene ID:466
- Modification:None
- Molecular Weight:29 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4°C
- Immunogen:This ATF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 193-219 amino acids from the C-terminal region of human ATF1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76071-346
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 193-219
Host: Rabbit
Isotype: Ig
Reactivity: Human