Anti-PSMD6 Rabbit Polyclonal Antibody
Catalog # 76066-120
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:26S proteasome non-ATPase regulatory subunit 6
- Antigen Symbol:PSMD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:154-182 aa
- Form:Liquid
- Gene ID:9861
- Modification:None
- Molecular Weight:46 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.36 mg/ml
- Shipping Temperature:4°C
- Immunogen:This PSMD6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 154-182 amino acids from the Central region of human PSMD6.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76066-120
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: PSMD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 154-182
Host: Rabbit
Isotype: Ig
Reactivity: Human