Anti-UCHL5 Rabbit Polyclonal Antibody
Catalog # 76065-978
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase isozyme L5
- Antigen Symbol:UCHL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:136-164 aa
- Form:Liquid
- Gene ID:51377
- Modification:None
- Molecular Weight:38 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This UCHL5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 136-164 amino acids from the Central region of human UCHL5.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76065-978
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: UCHL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 136-164
Host: Rabbit
Isotype: Ig
Reactivity: Human