Anti-CREB3 Rabbit Polyclonal Antibody
76065-948
: Prosci
- Antibody Type:Primary
- Antigen Name:Cyclic AMP-responsive element-binding protein 3
- Antigen Symbol:CREB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:66-94 aa
- Form:Liquid
- Gene ID:10488
- Modification:None
- Molecular Weight:44 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.38 mg/ml
- Shipping Temperature:4°C
- Immunogen:This CREB3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 66-94 amino acids from the N-terminal region of human CREB3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76065-948
For WB starting dilution is: 1:1000
Type: Primary
Antigen: CREB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 66-94
Host: Rabbit
Isotype: Ig
Reactivity: Human