Anti-CREB3L4 Rabbit Polyclonal Antibody
76069-536
: Prosci
- Antibody Type:Primary
- Antigen Name:Cyclic AMP-responsive element-binding protein 3-like protein 4
- Antigen Symbol:CREB3L4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 μL
- Epitope:338-366 aa
- Form:Liquid
- Gene ID:148327
- Modification:None
- Molecular Weight:43 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This CREB3L4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 338-366 amino acids from the C-terminal region of human CREB3L4.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76069-536
For WB starting dilution is: 1:1000
Type: Primary
Antigen: CREB3L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 338-366
Host: Rabbit
Isotype: Ig
Reactivity: Human