Anti-Cdk4 Rabbit Polyclonal Antibody
Catalog # 76068-768
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin-dependent kinase 4
- Antigen Symbol:Cdk4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 μL
- Epitope:219-246 aa
- Form:Liquid
- Gene ID:12567
- Modification:None
- Molecular Weight:34 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This Mouse Cdk4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 219-246 amino acids from the C-terminal region of mouse Cdk4.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76068-768
Specifications
About this item
For WB starting dilution is: 1:1000
Type: Primary
Antigen: Cdk4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 219-246
Host: Rabbit
Isotype: Ig
Reactivity: Mouse